Anti-ZNF124 Rabbit Polyclonal Antibody
10156-288
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 124
- Antigen Symbol:ZNF124
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:ZNF124
- Purification:Antigen affinity purification
- Cat. No.:10156-288
- Supplier no.:25216-1-AP
The ZNF124 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human ZNF124. This Antibody recognizes human Antigen. The ZNF124 Antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.
IHC 1 : 50 ; WB 1 : 600
Type: Primary
Antigen: ZNF124
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human