Anti-ZC3H12D Rabbit Polyclonal Antibody
Catalog # 10155-392
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc Finger CCCH-type Containing 12D
- Antigen Symbol:ZC3H12D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:MCP-induced protein 4|ZC3H12D|TFL|C6orf95|p34|Probable ribonuclease ZC3H12D|dJ281H8.1|MCPIP4
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:ZC3H12D
- Purification:Antigen affinity purification
- Cat. No.:10155-392
- Supplier no.:24991-1-AP
Specifications
About this item
The ZC3H12D antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZC3H12D. This antibody recognizes human antigen. The ZC3H12D antibody has been validated for the following applications: ELISA, IHC, WB analysis.
WB 1 : 600 ; IF 1 : 25 ; IHC 1 : 50
Type: Primary
Antigen: ZC3H12D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human