Order Entry
United States
ContactUsLinkComponent
2-Aminoisobutyric acid for synthesis, Sigma-Aldrich®
2-Aminoisobutyric acid for synthesis, Sigma-Aldrich®
Catalog # EM8.21244.0050
Supplier:  Avantor
CAS Number:  
2-Aminoisobutyric acid for synthesis, Sigma-Aldrich®
Catalog # EM8.21244.0050
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    50 g
  • Environmentally Preferable:
  • Cat. No.:
    EM8.21244.0050
  • Supplier no.:
    8.21244.0050

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    300 °C
  • Molar Mass:
    103.12 g/mol
  • Synonyms:
    α-Aminoisobutyric acid
  • Hill Formula:
    C4H9NO2
  • Chemical Formula:
    H2NC(CH3)2COOH

Specification Test Results

About this item

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid
 :
 :103.12 g/mol
CAS Number:62-57-7
Formula:C₄H₉NO₂
MDL Number:MFCD00008049
Boiling Pt:204
Melting Pt:335
Merck Index:13,00444
 :M-Clarity™ quality level = MQ200