Order Entry
United States
ContactUsLinkComponent
 
Oxidizing
Palladium(II) oxide Pd 85% for synthesis, hydrogenation catalyst, Sigma-Aldrich®
Catalog # EM8.07107.0001
Supplier:  MilliporeSigma
CAS Number:  
Oxidizing
Palladium(II) oxide Pd 85% for synthesis, hydrogenation catalyst, Sigma-Aldrich®
Catalog # EM8.07107.0001
Supplier:  MilliporeSigma
Supplier Number:  8.07107.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, pre-packed
  • Size:
    1 g
  • Cat. No.:
    EM8.07107.0001
  • Supplier no.:
    8.07107.0001

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    122.42 g/mol
  • Hill Formula:
    OPd
  • Chemical Formula:
    PdO

Specification Test Results

About this item

 :
 :122.42 g/mol
ADR:5.1,II
 :M-Clarity™ quality level = MQ200
UN:1479
 :GHS03
CAS Number:1314-08-5
Formula:OPd
MDL Number:MFCD00011172