4-Tolualdehyde for synthesis, Sigma-Aldrich®
Catalog # EM8.06179.0250
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:250 mL
- Cat. No.:EM8.06179.0250
- Supplier no.:8.06179.0250
Specifications
Specification Test Results
- Solubility:0.25 g/l (25 °C)
- Melting Point:-6 °C
- Molar Mass:120.15 g/mol
- Boiling Point:204 °C (1013 hPa)
- Vapor Pressure:0.33 hPa (25 °C)
- Flash Point:71.5 °C DIN 51758
- Refractive Index:1.5447 (20 °C, 589 nm)
- Explosion Limit:0.9 - 5.6 %(V)
- Density:1.02 g/cm3 (20 °C)
- Ignition Point:395 °C DIN 51794
- Synonyms:p-Tolualdehyde, p-Toluylaldehyde
- Hill Formula:C8H8O
- Chemical Formula:4-(CH3)C6H4CHO
Specification Test Results
About this item
:
:120.15 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:104-87-0
Formula:C₈H₈O
MDL Number:MFCD00006954