Molecular sieve 3A (0.3 nm, 3 Å), beads ~ 2 mm, Supelco®
Catalog # EM1.05704.0250
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:250 g
- Cat. No.:EM1.05704.0250
- Supplier no.:1.05704.0250
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Bulk Density:700 - 750 kg/m3
- Synonyms:Sodium aluminium silicate
Specification Test Results
About this item
:
CAS Number:308080-99-1
:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613