ContactUsLinkComponent
Feather A35 Low Profile Microtome Blades, Sakura
Feather A35 Low Profile Microtome Blades, Sakura
Catalog # 89199-474
Supplier:  Sakura Finetek
undefined
Feather A35 Low Profile Microtome Blades, Sakura
Catalog # 89199-474
Supplier:  Sakura Finetek
Supplier Number:  4980
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Feather A35 Low Profile Microtome Blades
  • Cat. No.:
    89199-474
  • Supplier no.:
    4980
Specifications
About this item
Low profile, ultra sharp blades section paraffin blocks without striations or chatter
Dispenser made to safely eject blades and store used blades. Blades are coated with a PTFE resin surface.
Ordering information: Blades are packaged with the dispenser. A case of 10 dispensers contains 50 blades per dispenser.