Anti-ADAM8 Rabbit Polyclonal Antibody
Catalog # 10155-132
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:ADAM Metallopeptidase Domain 8
- Antigen Symbol:ADAM8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:CD156a|CD156|MS2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:ADAM8
- Purification:Antigen affinity purification
- Cat. No.:10155-132
- Supplier no.:23778-1-AP
Specifications
About this item
The ADAM8 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ADAM8. This antibody recognizes human, mouse antigen. The ADAM8 antibody has been validated for the following applications: ELISA, WB, IHC analysis.
WB 1 : 300
Type: Primary
Antigen: ADAM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse