


TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) analytical reagent, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:9002-93-1
:9036-19-5
Formula:C₈H₁₇C₆H₄(OCH₂CH₂)⁷nOH
MDL Number:MFCD00132505
Merck Index:13,06793
Boiling Pt:>200 °C (1013 hPa)
Density:1.07 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Identity (IR spectrum):passes test
- pH value (10 g/l, H₂O, 20 °C):5.0 - 8.0
- Density (d 20/4):1.064 - 1.067
- Cloud point:63 - 69 °C
- Ethylene oxide (GC):≤30 ppm
- Dioxane (GC):≤20 ppm
- Sulphated ash (600 °C):≤0.2%
- Water (acc. to Karl Fischer):≤0.2%
Specification Test Results
Frequently Bought Together