Order Entry
United States
Orders LinkContactUsLinkComponent
StabilCoat® Plus Stabilizer, Surmodics

StabilCoat® Plus Stabilizer, Surmodics

50 mL
Catalog # 102867-010
Marketsource
Supplier:   Surmodics
Supplier #:  SC02-0050
StabilCoat® Plus Stabilizer, Surmodics
Catalog # 102867-010
Supplier:  Surmodics
Marketsource
CAS Number:  
StabilCoat® Plus Stabilizer, Surmodics

50 mL

CAS Number:  
Price
$40.00
Log in for availability
Each (50ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

StabilCoat® Plus Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilCoat® Plus stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.

  • 3 year shelf life
  • Increased signal
  • Reduced background noise
  • Increased sensitivity

StabilCoat® Plus microarray stabilizer improves the stability of proteins covalently bound to microspheres stored in solution and array proteins stored in the dried state.

Certifications: ISO 13485:2016 & 9001:2015 Certified

Specifications

Specifications

  • Size:
    50 mL
  • Cat. No.:
    102867-010
  • Storage Buffer:
    PBS
  • pH:
    7.0-7.4
  • Shelf Life:
    3 years
  • Protein:
    Bovine
  • Storage Temperature:
    2 to 8 °C or Room Temperature

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.