Order Entry
United States
Orders LinkContactUsLinkComponent
StabilZyme® SELECT Stabilizer, Surmodics

StabilZyme® SELECT Stabilizer, Surmodics

Marketsource
Supplier:   Surmodics
StabilZyme® SELECT Stabilizer, Surmodics
Supplier:  Surmodics
Marketsource
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The StabilZyme® SELECT Stabilizer is a multi-purpose stabilizing solution designed to maintain the conformation of antibody/antigen conjugates, antibody coated particles and unmodified proteins that are commonly used in immunoassays. Adding the conjugate/protein directly into the SELECT Stabilizer allows for stable storage of the conjugate at a working concentration and eliminates the need for subsequent dilutions.

  • Long-term in solution stability
  • longer shelf life for kits
  • 2.5 year shelf life

StabilZyme® SELECT Stabilizer is a general purpose in-solution protein stabilizer that can be used for HRP conjugates, standards and controls in immunoassay applications.

Certifications: ISO 13485:2016 & 9001:2015 Certified

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 102466-328
  • 102466-330
  • Application:
  • ELISA, Western Blot, ImmunoBlot
  • ELISA, Western Blot, ImmunoBlot
  • pH:
  • 6.5-7.5
  • 6.5-7.5
  • Protein:
  • Bovine
  • Bovine
  • Storage Buffer:
  • None
  • None
  • Storage Temperature:
  • 2 to 8 °C or Room Temperature
  • 2 to 8 °C or Room Temperature
  • Shelf Life:
  • 2.5 years
  • 2.5 years

Specifications

Product Family Options

Product Information

  • Conjugate
    Size
    Availability
    Price
  • 102466-328
    StabilZyme SELECT® Stabilizer
    StabilZyme SELECT® Stabilizer
    HRP/General
    50 mL
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $40.00
    $40.00
    Each (50ml)
       $40.00

    Supplier Number: SZ03-0050
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102466-328
    Application
    ELISA, Western Blot, ImmunoBlot
    pH
    6.5-7.5
    Protein
    Bovine
    Storage Buffer
    None
    Storage Temperature
    2 to 8 °C or Room Temperature
    Shelf Life
    2.5 years
    Marketsource
  • 102466-330
    StabilZyme SELECT® Stabilizer
    StabilZyme SELECT® Stabilizer
    HRP/General
    1 L
    Log in for availability
    Log in for availability
    Each (1,000ml)
    Each (1,000ml) - $240.00
    $240.00
    Each (1,000ml)
       $240.00

    Supplier Number: SZ03-1000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102466-330
    Application
    ELISA, Western Blot, ImmunoBlot
    pH
    6.5-7.5
    Protein
    Bovine
    Storage Buffer
    None
    Storage Temperature
    2 to 8 °C or Room Temperature
    Shelf Life
    2.5 years
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.