About this item
StabilZyme® HRP, StabilZyme® Protein-Free and StabilZyme® NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
- Long Term in-solution stability
- Increased shelf life of kits
- 2.5 year stability
StabilZyme® HRP Conjugate Stabilizer is specifically optimized for the HRP enzyme and is the gold standard for in-solution conjugate stability.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein:Bovine
- Shelf Life:2.5 years
- Storage Buffer:MOPS
- Storage Temperature:2 to 8 °C or Room Temperature
- pH:6.2-6.7