MAbPac™ HIC-Butyl HPLC Columns, Thermo Scientific
The Thermo Scientific MAbPac™ HIC-Butyl column is a high-resolution polymer-based hydrophobic interaction chromatography (HIC) column designed for the separation of mAbs and antibody-drug conjugates (ADCs). The hydrophilic resin and optimal density of the butyl groups lead to excellent bio-compatibility and low carryover. In addition, MAbPac HIC-Butyl column provides complementary selectivity to MAbPac HIC-10 and MAbPac HIC-20 columns.
- Optimal selectivity for ADCs
- High resolution and high efficiency
- Stable at wide range of pH (2−12)
- Low carryover
- Rugged column packing
The MAbPac HIC-Butyl column is based on hydrophilic, non-porous, 5 μm polymer particles functionalized with butyl groups. The hydrophilic resin provides biocompatibility and low carryover while the optimal density of the butyl groups provides high resolution separation.
: Guard cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2.
- Catalog No:
- 10754-142
- 10754-140
- Description:
- 10754-142
- 10754-140
- pH Range:
- 2 to 8
- 2 to 8
Frequently Bought Together