Order Entry
United States
ContactUsLinkComponent
 
Tissue-Tek Rapid Tissue Processors, Sakura
Catalog # 89199-776
Supplier:  Sakura Finetek
CSA Certified
UL Listed
undefined
Tissue-Tek Rapid Tissue Processors, Sakura
Catalog # 89199-776
Supplier:  Sakura Finetek
Supplier Number:  7720
CSA Certified
UL Listed

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Power:
    Single Phase, NEMA L6-20 plug
  • Description:
    Tissue-Tek X120 Rapid Tissue Processor
  • Cat. No.:
    89199-776
  • W×D×H:
    170 x 71 x 162 cm
  • Electrical:
    200 VAC, 50/60 Hz, 15 A
  • Supplier no.:
    7720

Specifications

About this item

Continuous tissue processing utilizing low-wattage microwave technology, molecular-friendly reagents and vacuum infiltration for rapid turn around.

  • Throughput of up to 120 specimens per hour, loading 40 cassettes every 20 minutes (X120) and up to 45 specimens every 45 minutes (X50)
  • Biopsy and larger specimens can be processed in same run

2 USB ports, external alarm output, UPS signalinput, LAN connection

Ordering information: No formalin or xylene is used on the X120 processor.