About this item
Providing a whole new level of light-duty comfort and performance, nitrile gloves feature the coolness and comfort of cotton, but with far superior grip.
- Superior grip compared to cotton gloves
- Resistant to snags, punctures, and abrasions
- Repellent properties against oil and grease
These gloves make it easier and safer to handle smooth objects like boxes or appliances.
Use in the automotive and transportation, chemical, glass, machinery and equipment, and metal fabrication industries. Uses include appliance manufacture; component assembly; packaging; shipping and receiving; textile manufacture: threads, fabrics, mattresses, and cables; transmission, motor assembly, packaging, and inspection; and warehouse work.
Coated only on fingers and palm, gloves protect the working area, but leave the back of the hand free to breathe. Gloves provide resistance to snags, puncture and abrasion. Nitrile coating repels oil and grease. Gloves last longer than an ordinary cotton glove.
Specifications
- Catalog No:
- 32889-794
- 32889-798
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Liner Material:
- 100% Cotton
- 100% Cotton
- Finish:
- Dipped/Lightly Textured
- Dipped/Lightly Textured
- Cuff Style:
- Knit Wrist
- Knit Wrist
- EN Norm:
- 2121
- 2121