Hynit® 32-125 Nitrile Gloves with Slip-On Cuff, Perforated Back, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
A light, comfortable and longer-lasting replacement for cotton and leather, this special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility.
- Comfortable and long-lasting
- Excellent abrasion resistance
A liquid-repellent impregnation repels oil, grease, and dirt.
Use in the automotive and transportation, building and construction, logistics and warehousing, machinery and equipment, maintenance, metal fabrication, public utilities, and recycling industries. Uses include assembly of white goods; component assembly; light material handling; packaging; shipping and receiving; steel mill general handling; and textile manufacture: threads, fabrics, mattresses, and cables.
Certifications: ANSI level 1 cut protection; ANSI Level 3 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Specifications
- Catalog No:
- 32889-924
- 32889-914
- 32889-922
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Liner Material:
- 100% Cotton
- 100% Cotton
- 100% Cotton
- Finish:
- Fully Impregnated/Perforated Back
- Fully Impregnated/Perforated Back
- Fully Impregnated/Perforated Back
- Cuff Style:
- Slip-On
- Slip-On
- Slip-On
- EN Norm:
- 3111
- 3111
- 3111