Dip & Spin® Urine Dipstick and Microscopics Controls, Quantimetrix
Catalog # 82024-122
Supplier: Quantimetrix
CAS Number:
Specifications
- Description:Dip & Spin® Urine Dipstick and Microscopics Control
- Cat. No.:82024-122
- Volume:4× 120 ml
- Supplier no.:1470-01
Specifications
About this item
Dip & Spin is our most complete urinalysis control – two controls in one easy-to-use squeeze bottle for simple dispensing. We include values for microscopic measurements and all dipstick analytes plus microalbumin and creatinine, and qualitative results for hCG early pregnancy detection test methods.
- Two controls in one
- Liquid, made with human urine
- 18-month shelf life and open vial stability from date of manufacture when stored at 2 to 8°C
Dip&Spin is designed for use with most urinalysis reagent strips and standardized urinalysis systems and analyzers. It can also be used for confirmatory tests and refractometry.
Ordering information: Box includes 2 bottles each of Level 1 and 2. Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.