Dropper® Plus Point-of-Care Urine Dipstick Controls, Quantimetrix
82024-130
:
- Description:Dropper® Plus POC Urine Dipstick Control
- Cat. No.:82024-130
- Volume:10× 5 ml
- Includes:Level 1 and 2
- Supplier no.:1440-04
Dropper Plus POC Urine Dipstick Control eliminates storage problems and provides maximum portability by providing one month of room temperature stability – perfect for sites without refrigeration. Easy-to-use dropper bottles make dispensing simple. We include values for all dipstick analytes plus microalbumin and creatinine; and qualitative results for hCG early pregnancy detection test methods. Dropper Plus POC Urine Dipstick Control is designed for use with most urinalysis reagent strips. It can also be used for confirmatory tests and refractometry.
- Liquid, made with human urine
- 1-month open vial stability at room temperature for maximum portability
- 18-month shelf life and open vial stability from date of manufacture when stored at 2 to 8°C
: Box includes 5 vials each of Level 1 and 2. Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.