Tris (1 M) Buffer Solution, pH 9.8, Quality Biological
Catalog # 10128-572
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Tris, 1 M
- Size:1 L
- Environmentally Preferable:
- Sterility:Sterile filtered
- Test Description:DNase, RNase and protease tested
- Shelf Life:36 months
- Shipping Temperature:Ambient
- Cat. No.:10128-572
- pH:9.8
- Supplier no.:351-176-131
Specifications
About this item
Suitable for molecular biology, protein chemistry, and biochemical applications.
- DNase, RNnase and Protease tested.
- Filtered through a 0.22 µm membrane
Tris Buffer, 1M pH 9.8 is commonly used in SDS-PAGE and electrophoresis applications.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: This product requires special production. Please allow 2 to 4 weeks for delivery.
Caution: Research use only.