TBS HCl (10X) Buffer Solution, pH 7.2, Quality Biological
Catalog # 10128-500
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:TBS HCl (10X), pH 7.2
- Size:500 ml
- Environmentally Preferable:
- Sterility:Sterile filtered
- Test Description:DNAse, RNAse and protease tested
- Shelf Life:36 months
- Shipping Temperature:Ambient
- Cat. No.:10128-500
- pH:7.2
- Supplier no.:351-058-101
Specifications
About this item
Tris HCl, 10X pH 7.2 is a Buffered Saline Solution used in procedures for the isolation of DNA.
- Filtered through a 0.22 µm membrane
- DNase, RNase, and Protease tested
Tris buffered saline is useful for the following: as a biological diluent buffer, as a buffer for antibodies, as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.