YPG Plates, Quality Biological
YPG Plates, 100× 30 ml, 25 mL
Catalog # 10128-348
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier: QUALITY BIOLOGICAL, INC.
Supplier #: 340-190-301
YPG Plates, Quality Biological
Catalog # 10128-348
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
YPG Agar is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose.
- YPG is a complex medium containing a nonfermentable carbon source (glycerol) that does not support the growth of ρ– or pet mutants.
YPG Plates are useful for isolation of Beta Carotene-Hyper producing Mutants and biocatalyst preparation.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Specifications
Specifications
- Description:YPG Plates
- Size:25 mL
- Volume:100× 30 ml
- Storage Conditions:2°C to 8°C
- Cat. No.:10128-348
- Sterility:Autoclaved
- Shipping Conditions:Ambient Temperature



