ContactUsLinkComponent
U-Channel Sign Post Driver, Brady
U-Channel Sign Post Driver, Brady
Catalog # 89007-850
Supplier:  Brady Worldwide
undefined
U-Channel Sign Post Driver, Brady
Catalog # 89007-850
Supplier:  Brady Worldwide
Supplier Number:  87982
Specifications
  • Description:
    U-Channel Sign Post Driver
  • Cat. No.:
    89007-850
  • Supplier no.:
    87982
Specifications
About this item
Brady's U-Channel Sign Post Driver makes it easy to install U-Channel sign posts
U-Channel sign posts are ideal for traffic signs and utility signage; this post driver will help you ensure that they are installed correctly. The U-Channel Sign Post Driver is made of heavy duty steel with black finish to withstand harsh conditions. It is 30" long and 4" diameter.