Dry Swabs, Sterile, Starswabs II™, Starplex Scientific
89413-034
: Starplex
- Description:Dry swabs, in tube
- Length:15.2 cm
- Cat. No.:89413-034
- Supplier no.:SO9D
Dry, sterile swabs with plastic shafts. Ideal for assessing microbial presence on various surfaces and equipment.
- Swab has a white plug cap
- Round Bottom tube labeled with lot number and expiration date
- Thick wall prevents dehydration
Rayon swab tip is inert, non-toxic and permits good sample retrieval and absorption.
: Swabs are packaged in round bottom tubes, 500 per case.