ContactUsLinkComponent
Original Cable Lockout, Brady Worldwide®
Original Cable Lockout, Brady Worldwide®
Catalog # 12222-800
Supplier:  Brady Worldwide
undefined
Original Cable Lockout, Brady Worldwide®
Catalog # 12222-800
Supplier:  Brady Worldwide
Supplier Number:  65318
Specifications
  • Color:
    Red
  • Description:
    Original Cable Lockout
  • Material:
    Impact Modified Nylon
  • Size:
    6' L x 1/8" Dia.
  • Cat. No.:
    12222-800
  • Supplier no.:
    65318
Specifications
About this item
The original Brady® cable lockout device is ideal for difficult lockout situations, such as large gate valves or oversized devices
Simply squeeze device to tighten cable, and insert padlock to secure the handle. Body is made of impact modified nylon, available in red, blue, or yellow. Standard lockout device is supplied with cable. Additional 6', 8', and 10' cables are sold separately.