Extra-Large Enclosed Lockout Station (Station Only), Brady Worldwide®
Catalog # 89007-240
Supplier: Brady Worldwide
Specifications
- Color:Red on White
- Description:Extra-Large Enclosed Lockout Station (Station Only)
- Material:Polystyrene
- Size:30"H x 25"W x 5.25"D
- Cat. No.:89007-240
- Supplier no.:50995
Specifications
About this item
Constructed of sturdy white polystyrene with predrilled holes for easy wall mounting
Pegboard-style holes for hooks are drilled into the back (8 hooks supplied standard). Built-in lock rail holds 20 to 25 locks, depending on lock size. Components sold seperately.