Extra-Large Lockout Station (Station Only), Brady Worldwide®
Catalog # 89007-236
Supplier: Brady Worldwide
Specifications
- Color:Red on White
- Description:Extra-large lockout station (station only)
- Material:Polystyrene
- Size:30"Hx25"Wx5.25"D
- Cat. No.:89007-236
- Supplier no.:50993
Specifications
About this item
Constructed of sturdy white polystyrene with predrilled holes for easy wall mounting.
- Pegboard-style holes for hooks are drilled into the back (8 hooks supplied standard)
- Built-in lock rail holds 20 to 25 locks, depending on lock size
- Components sold seperately