Lockout for Miniature SMC Models AR2000/NAR2000, Brady Worldwide®
Catalog # 12222-826
Supplier: Brady Worldwide
Specifications
- Description:Lockout for Miniature SMC Models AR2000/NAR2000
- Cat. No.:12222-826
- Supplier no.:64539
Specifications
About this item
Lockout enables regulator to be locked in the on, off, or throttled position
Easy installation involves removing snap cap, installing lockout on regulator rod, and snapping replacement cap on regulator. Supplied with lockout, cap, and allen wrench.