ContactUsLinkComponent
Lockout for Intermediate SMC Models AR3000/NAR3000, Brady Worldwide®
Lockout for Intermediate SMC Models AR3000/NAR3000, Brady Worldwide®
Catalog # 12222-828
Supplier:  Brady Worldwide
undefined
Lockout for Intermediate SMC Models AR3000/NAR3000, Brady Worldwide®
Catalog # 12222-828
Supplier:  Brady Worldwide
Supplier Number:  64540
Specifications
  • Description:
    Lockout for Intermediate SMC Models AR3000/NAR3000
  • Cat. No.:
    12222-828
  • Supplier no.:
    64540
Specifications
About this item
Lockout enables regulator to be locked in the on, off, or throttled position
Easy installation involves removing snap cap, installing lockout on regulator rod, and snapping replacement cap on regulator. Supplied with lockout, cap, and allen wrench.