Order Entry
United States
Orders LinkContactUsLinkComponent
Formalin Neutralizing Pads, 11"×10"

Formalin Neutralizing Pads, 11"×10"

Formalin neutralizing pads, 11×10", Roll of 55
Catalog # 470136-436
Supplier #:  ES728
Formalin Neutralizing Pads, 11"×10"
Catalog # 470136-436
Formalin Neutralizing Pads, 11"×10"
Formalin Neutralizing Pads, 11"×10"

Formalin neutralizing pads, 11×10", Roll of 55

Price
$66.95
Log in for availability
Roll of 55 Case of 330
Product Details & Documents

Create a safer and more enjoyable dissection experience with these odor-absorbing pads, designed to minimize exposure to formaldehyde off-gassing and neutralize excess formalin.

  • Removes odor
  • Decrease distraction

Create a safer and more enjoyable dissection experience with these odor-absorbent pads, designed to minimize exposure to formaldehyde off-gassing and neutralize excess formalin. Simply line your specimen pan with a pad and place the specimen on top to immediately absorb excess formalin and odors during specimen dissection.

Specifications
  • Description:
    Formalin neutralizing pads
  • Size:
    11×10"
  • Packaging:
    Roll of 55
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.