T100™ Thermal Cycler, Bio-Rad
Product Details & Documents
The compact T100™ Thermal Cycler is a small footprint personal thermal cycler
- Easy intuitive programming and use, with a color touch screen
- Thermal gradient technology can optimize an assay in a single run
- Store up to 500 protocols, and an unlimited number on USB drives
- Built-in library of protocols including long PCR, fast PCR and RT-PCR
- Small footprint
- Fits in any laboratory
Thermal gradient technology permits quick reaction optimization in a single run by simultaneously testing eight different temperatures across a range of up to 25°C. The cycler has graphical programming and a library of standard PCR protocols. Easy protocol management with personalized folders or a USB flash drive, which can also be used for protocol transfer and backup.
Specifications
- Description:T100™ Thermal Cycler
- Electrical:100 - 150 VAC, 50/60 Hz; 220 - 240 VAC, 50/60 Hz; 700 W max.
- Details:Thermal Cycler, Power Cord and Tube Support Ring
- Programs:500 typical programs; unlimited with USB flash drive expansion
- Gradient:+
- Temperature Accuracy:±0.5°C of programmed target
- Heating Ramp Rate:2.5 °C/s
- Weight:9 kg (20 lbs.)
- Interfaces:1 USB A
- Cat. No.:100446-370
- Gradient Temperature Range:30–100 °C (±0.5 °C of programmed temperature)
- Sample capacity:96 x 0.2 mL tubes, 0.2 mL Tube Strips or 1 x 96-Well Plate
- Temperature Duration Accuracy:1–25°C
- Temperature Range:4–100°C
- W×D×H:26×47×23 cm (10×18×9")
- Temperature Uniformity:±0.5°C well-to-well within 30 sec of arrival at target temperature
- Display:5.7'' VGA Color Touch Ccreen
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tube Support Ring for Extra Support, For Use With Individual Tubes, Pack of 2Tube Support Ring for Extra Support, For Use With Individual Tubes, Pack of 2100446-372Log in for availabilityEach - $60.00
Product Information
- Catalog #AvailabilityPrice
- Touch-Screen Protector to Shield From Dust, Reagents and Scratches, Clear Plastic, Pack of 2Touch-Screen Protector to Shield From Dust, Reagents and Scratches, Clear Plastic, Pack of 2100446-374Log in for availabilityEach - $69.23
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



