ContactUsLinkComponent
iQ™ 96-Well Real-Time PCR Plates, High Profile, Bio-Rad
Catalog # 101645-684
Supplier:  Bio-Rad
iQ™ 96-Well Real-Time PCR Plates, High Profile, Bio-Rad
Catalog # 101645-684
Supplier:  Bio-Rad
Supplier Number:  2239441
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • No. of wells:
    96
  • Profile:
    High
  • Skirt:
    Semi-Skirted
  • Well volume:
    0,33 ml
  • Cat. No.:
    101645-684
  • Supplier no.:
    2239441
Specifications
About this item
iQ™ 96-Well Real-Time PCR Plates are high-profile, semi-skirted plates, compatible with many real-time PCR detection systems including, the Bio-Rad 1000 series, Applied Biosystems standard systems, the Eppendorf Mastercycler series and Agilent (Stratagene) MX series.
  • Half-height skirt adds stiffness and a labeling surface
  • Recommended volumes of 5–125 µL (330 µL maximum)
  • Perforated every three columns for easy setup of triplicate reactions
  • Free of DNase, RNase, and human DNA