ContactUsLinkComponent
Precision Plus Protein™ Prestained Standards, Protein Markers, Bio-Rad
 :  Bio-Rad
Precision Plus Protein™ Prestained Standards, Protein Markers, Bio-Rad
 :  Bio-Rad
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Storage Conditions:
    Store standards for 6 months at 4°C or –20°C until expiration date; store Kaleidescope standards for 12 months at –20°C.
  • Loading Volume:
    Electrophoresis: 10 µL; Blotting: 7 µL; Fluorescent Blot Detection: 5 µL
  • Prestained Protein Markers:
    Blue/635 nm laser (red LED); Pink/532 nm laser (green LED); Green/635 nm laser (red LED); Yellow/488 nm laser (blue LED)
  • Type:
    Covalently stained with multicolored dyes
Frequently Bought Together