ContactUsLinkComponent
Accessories for UniSampler®, Sampler with Hose, Bürkle
Accessories for UniSampler®, Sampler with Hose, Bürkle
Catalog # 10217-064
Supplier:  BURKLE INC
undefined
Accessories for UniSampler®, Sampler with Hose, Bürkle
Catalog # 10217-064
Supplier:  BURKLE INC
Supplier Number:  5314-2002
Specifications
  • Description:
    Suction hose for replacement PVC, 2.5 m
  • Cat. No.:
    10217-064
  • Supplier no.:
    5314-2002