LaboPlast® Stopcock PA, Electroconductive, Bürkle
Catalog # 10216-912
Supplier: BURKLE INC
Specifications
- Description:Spigot PA, electroconductive, 3/4" inner thread nut
- Cat. No.:10216-912
- Tubing needed, I.D.:¾" inflowing side, spout/nozzle on outflowing side
- Supplier no.:0525-1100
Specifications
About this item
Stopcock with tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm.
- With discharge angle and rotatable spout preventing drips
- Media-contacting parts PA conductive to electricity and PTFE
- Supplied with PA adapter conductive to electricity for 3/4" inner thread
Stopcock PA made of electrically conductive polyamide, suitable for solvents.