Cold Extractor, COEX, VELP Scientifica
About this item
Cold extractor that performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE Series for raw fiber analysis.
- Precise Results
- Versatile
- Convenient
Precise Results–Reduces fat to below 1% improving analytical precision. No loss – reduced sample remains in same crucible for analysis.
Versatile–Immediate raw fiber extraction after fat extraction here. Processing of individual or several samples.
Convenient–Fatty components collected for easy disposal. All reagents can be added through the same channel.
Ordering information: Combine with FIWE for raw fiber analyzer.
1 Options Available Below
Product Details & Documents
Cold extractor that performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE Series for raw fiber analysis.
- Precise Results
- Versatile
- Convenient
Precise Results–Reduces fat to below 1% improving analytical precision. No loss – reduced sample remains in same crucible for analysis.
Versatile–Immediate raw fiber extraction after fat extraction here. Processing of individual or several samples.
Convenient–Fatty components collected for easy disposal. All reagents can be added through the same channel.
Ordering information: Combine with FIWE for raw fiber analyzer.
Specifications for Product Family
Specifications
- Catalog No:
- 10184-186
- 10184-180
- 10184-174
- Weight:
- 19 kg
- 19 kg
- 19 kg
- Power:
- 120W
- 120W
- 120W
- W×H×D:
- 730×300×380 mm
- 730×300×380 mm
- 730×300×380 mm
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 10184-186COEX Cold Extractor, 115V/60 HzSupplier Number: F30540204Specifications:
More
Specifications
Cat. No.10184-186Weight19 kgPower120WW×H×D730×300×380 mm - 10184-180COEX Cold Extractor, 230V/60 HzCOEX Cold Extractor, 230V/60 HzDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.10184-180Weight19 kgPower120WW×H×D730×300×380 mm - 10184-174COEX Cold Extractor, 230V/50 HzCOEX Cold Extractor, 230V/50 HzDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.10184-174Weight19 kgPower120WW×H×D730×300×380 mm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



