ContactUsLinkComponent
Nanoject II™ Microinjectors, Drummond Scientific
Nanoject II™ Microinjectors, Drummond Scientific
Catalog # 490007-164
Supplier:  Avantor
undefined
Nanoject II™ Microinjectors, Drummond Scientific
Catalog # 490007-164
Supplier:  Avantor
Specifications
  • Description:
    Nanoject II Variable Volume (2.3 to 69 nL), Automatic Injector with Glass Capillaries (110V)
  • Environmentally Preferable:
  • Cat. No.:
    490007-164
  • Supplier no.:
    3-000-204
Specifications
About this item
The Nanoject II Auto-Nanoliter injector performs ultra-delicate nanoliter injection procedures into oocytes, embryos, and tissues.
  • Automated, Microprocessor-Controlled Microinjection Pipet Delivers Precise Nanoliter Volumes
  • Remote Control and Non-Rotating Plunger Eliminate Vibration for Precise Injection
  • Separate Foot Switch Available for Hands-Free Operation
  • Mountable on Most Micromanipulators
  • Supplied with Enough Borosilicate Glass Capillaries to Prepare 400 Injection Tips
The automated, microprocessor-controlled microinjection pipet delivers precise nanoliter volumes
The injector can be mounted on most micromanipulators and includes enough glass capillaries to prepare 400 injection tips with a standard softening point of 780°C.
The remote control and non-rotating plunger eliminate vibration for precise injection, minimizing possible cell rupture.