About this item
Readily soluble in water.
One unit will liberate one µmole of glucose from cellulose in 1 hr at pH 5 and 37 °C. ; CMC Unit Definition: 100 units is sufficient to produce reducing sugar equivalent to 1 mg of glucose per minute at pH 5.0 and 40 °C.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:9012-54-8
- Source:Trichoderma virde
- Storage Conditions:Store at −0 °C.
- Form:Tan-free flowing powder
- Enzyme Name:Cellulase
- Enzyme Synonyms:1,4-(1,3:1,4)-β-D-Glucan 4-glucano-hydrolase, E.C. 3.2.1.4
- pH:Optimum pH: Varies with the substrate in the range 4.2 - 5.2.(Lit.)