About this item
Pyruvate kinase from rabbit muscle is a tetramer consisting of four equal subunits of molecular weight 57 kDa. Pyruvate kinase catalyzes the following reaction: ADP + Phosphoenolpyruvate → Pyruvate + ATP . Both Mg2+ and K+ are required metal cofactors for optimal activity. The enzyme is inhibited by Ca2+. It is a key enzyme in glycogen metabolism.
- Presentation: White Crystalline Powder
- Isoelectric Point (pl): 7.6
- Optimum pH: 7.5
- Dissolves readily at 5 mg/mL in 0.06M Tris/HCI pH 7.4 to give a clear colorless solution.
Pyruvate kinase is often used as a coupling enzyme in conjunction with lactic dehydrogenase in quantifying ADP and the activity of enzymes that catalyze the formation of ADP.
Pyruvate kinase is highly specific for phosphoenolpyruvate, but can utilize other dinucleotide triphosphates as substrates in place of ATP including GTP, ITP, dATP, UTP, and CTP.
Protein determined by biuret.
That amount of enzyme causing the oxidation of one micromole of NADH per minute at 25 °C and pH 7.4.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:9001-59-6
- Source:Muscle
- Species:Rabbit
- Storage Conditions:Store at -20 °C, Desiccated.
- Form:White Crystalline Powder
- Enzyme Name:Pyruvate Kinase
- Enzyme Activity:≥200 U/mg protein