About this item
Glucose-6-phosphate dehydrogenase (G6PD or G6PDH) is a cytosolic enzyme in the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH).
Contaminants: CPK, 6-PGDH, PGI, PGM: < 0.01% HK: < 0.05%
Either NAD+ or NADP+ serves as coenzyme, the reaction velocity with NAD+ being approximately 1.8 times greater than with NADP+ p-Glucose-6-phosphate is a preferential substrate for the enzyme, although p-Glucose reacts slowly, Fructose-6-phosphate, fructose-1, 6-diphoshate and ribose-5-phosphate are not considered to be substrates.
Unit definition: One unit will reduce 1.0 µmole of NAD+ per minute at pH 7.8 and 25°C.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:9001-40-5
- Species:Leuconostoc mesenteroides
- Storage Conditions:−20 °C
- Form:White lyophilized powder
- Enzyme Name:Glucose-6-Phosphate Dehydrogenase
- Enzyme Activity:≥400 u/mg solid
- Solubility:Dissolves in 0.1 M TEA/NaOH pH 8.0 (5 mg/ ml-clear, colorless solution), also in distilled water or dilute buffer
- Molecular Weight:104 kDa
- pH:5.5 - 7.5