About this item
Casein hydrolysate is a hydrochloric acid digest of casein.
- Protein molecule has been substantially reduced to its constituent amino acids
- Protein hydrolysate has been specially processed to remove essentially all sodium chloride
- Soluble in water (2%), no haziness after autoclaving
- FTIR: Conforms to Reference
- ≥55% Light Transmission at 450nm
- Water Content: <3%
Laboratory media including basal media for vitamin analysis of folic acid, pantothenic acid, riboflavin, thiamin, niacin, and B-12. It is also an inexpensive source of mixed amino acids.
Laboratory media including basal media for vitamin analysis of folic acid, pantothenic acid, riboflavin, thiamin, niacin, and B-12. It is also an inexpensive source of mixed amino acids.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Conditions:Store at Room Temperature (15-30 °C).
- Form:Off-white Powder
- Protein/Peptide Name:Casein Hydrolysate
- pH:6.0 - 7.0 (2% solution)