About this item
Lipases are a subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
- Presentation: Cream Colored Powder
- Extinction Coefficient (E1%): E1%280 = 13.3(Lit.)
- Isoelectric point (pI): Lipase A = 4.9 and Lipase B = 5.0 (Lit.)
- Solubility: Partly soluble in water and insoluble in alcohol.
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions.They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
One USP unit of lipase activity is contained in the amount of pancreatin that liberates 1.0 µEq of acid per minute at a pH of 9.0 at 37 °C under the conditions of the assay for lipase activity.
Lipases are a subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: