About this item
Soluble in water
One unit is defined as the amount of enzyme that will liquefy a standard clot of fibrinogen, plasminogen and thrombin in 10 minutes at 37 °C and pH 7.
Streptokinase catalyzes hydrolysis of amide linkages. Forms a 1:1 complex with plasminogen which is then converted to streptokinase-plasmin. Both streptokinase-plasminogen and streptokinase-plasmin complexes can activate plasminogen. As a result of its interaction with human or rabbit plasminogen, streptokinase undergoes specific fragmentation.
Streptokinase is commonly used as a thrombolytic agent in the therapy of ischemic stroke. This therapy carries the important risk of intracerebral hemorrhage. Streptokinase is also used in the treatment of complicated parapneumonic effusions and empyema where adverse reactions, allergic type, are rare
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:9002-01-11
- Form:Lyophilized solid
- Enzyme Name:Streptokinase