Soluble in water; Insoluble in ethanol, acetone and isopropyl alcohol.
One unit will hydrolyze 1.0 µmole of o-nitrophenyl-beta-D-galactoside to o-nitrophenol per minute at pH 4.5 and 30 °C.
Stability: Stable in pH range of 4.0 to 8.0. Stable to heat. Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate, respectively(Lit.)
Lactase is an enzyme preparation produced by Aspergillus oryzae fermentation. Lactase hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and a-D-Glucose.
Lactase is used to screen for blue white colonies in the MCS of various plasmid vectors in Escherichia coli or other bacteria. Its primary commercial use is to break down lactose in milk to make it suitable for people with lactose intolerance.
β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Source:Aspergillus oryzae
- Storage Conditions:Store at +4 °C.
- Form:Off-white powder
- Enzyme Name:Lactase
- Enzyme Activity:>5000 U/g
- Enzyme Synonyms:β-D-Galactoside galactohydrolase, Lactase from Aspergillus oryzae, β-Galactosidase, E.C. 3.2.1.23