Deoxyribonuclease from beef pancreas, DNase I, was first crystallized by Kunitz. It is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding 5'-phosphate terminated polynucleotides with a free hydroxyl group on position 3'. The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.
- Presentation: White Powder
- Extinction Coefficient (E1%): E1%280= 11.1
- A lyophilized powder containing a small amount of glycine-stabilizer
- Optimum pH: 7.8
DNase I is used as a genetic marker for forensic identification. Used for the removal of DNA from protein samples. DNAse I is used to nick DNA, as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used in the nuclease stock along with RNAse during cell lysate preparation of Madin-Darby canine kidney (MDCK) II cell lines. It has also been used during RNA extraction from Sindbis virus. Deoxyribonuclease I from bovine pancreas has been used for the identification, localization and expression of two novel human genes similar to deoxyribonuclease I. Deoxyribonuclease I from bovine pancreas has also been used in a study to investigate a new approach to obtaining high-activity RNase, DNase, cholesterolesterase, and trypsin from cattle pancreas.
DNase I is an endonuclease that acts on phosphodiester bonds adjacent to pyrimidines to produce polynucleotides with terminal 5′-phosphates. In the presence of Mg²⁺, DNAse I cleaves each strand of DNA independently and the cleavage sites are random. Both DNA strands are cleaved at approximately the same site in the presence of Mn2+. The pH optimum is found to be between 7 and 8. Divalent cations such as Mn²⁺, Ca²⁺, Co²⁺, and Zn²⁺ are activators of the enzyme. A concentration of 5 mM Ca²⁺ stabilizes the enzyme against proteolytic digestion. DNAse I from bovine pancreas consists of four chromatographically distinguishable components, A, B, C, and D, with their molar ratios being 4:1:1 respectively. Only minor amounts of D are found. 2-Mercaptoethanol, chelators, sodium dodecyl sulfate (SDS) and actin are known to inhibit the enzyme activity.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:9003-98-9
- Source:Pancreas
- Species:Bovine
- Storage Conditions:Store at +4 °C. Store Desiccated.
- Form:white powder
- Enzyme Name:Deoxyribonuclease I
- Enzyme Activity:≥2000 kunitz U/mg dw
- Enzyme Synonyms:DNase I, Deoxyribonucleate 5′-oligonucleotido-hydrolase
- Formulation:A lyophilized powder containing a small amount of glycine-stabilizer