Orders LinkContactUsLinkComponent
Space Drape, AIMS™
Space Drape, AIMS™
Warming Drape
Catalog # 95059-642
Supplier:   Aims
Supplier #:  SPDR
Space Drape, AIMS™
Catalog # 95059-642
Supplier:  Aims
Space Drape, AIMS™
Space Drape, AIMS™
Warming Drape
Price
$20.45
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The space drape is designd to keep lab animals warm after the completion of procedures.
It features a soft flannel-like material on one side and a reflective coating on the other side.
Specifications
  • Description:
    Warming Drape