Order Entry
United States
ContactUsLinkComponent
 
Snap Plug Caps For 8×40 mm Shell Vials, DWK Life Sciences
Catalog # 66012-310
Small Business Enterprise
undefined
Snap Plug Caps For 8×40 mm Shell Vials, DWK Life Sciences
Catalog # 66012-310
Supplier Number:  242800
Small Business Enterprise

Specifications

  • Cap Size:
    8 mm
  • Description:
    Snap Plug Cap
  • Cat. No.:
    66012-310
  • Cap color:
    Natural
  • Supplier no.:
    242800

Specifications

About this item

Polyethylene snap plug cap.

  • Snap plug closures available in 5 colors
  • Purchase vials and caps separately or together in convenience packs