Order Entry
United States
Orders LinkContactUsLinkComponent
Pipemidic acid 98%

Pipemidic acid 98%

Pipemidic acid 98%
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Pipemidic acid, 98%

Purity: 98% CAS No.: 51940-44-4 Molecular Formula: C14H17N5O3 Formula Weight: 303.32 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Sensitivity: Light Sensitive Appearance: Off-white Melting Point: 251-255° Boiling Point: 717° Solubility: Soluble in 1N sodium hydroxide or in DMSO. Insoluble in water MDL No.: MFCD00057291

Synonyms:  8-Ethyl-5-oxo-2-(piperazin-1-yl)-5,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylic acid
  • CAS Number:51940-44-4
  • Formula:C₁₄H₁₇N₅O₃
  • MDL Number:MFCD00057291
  • Molecular Weight:303.322 g/mol
  • Merck Index:14,7460
  • Boiling Pt:717 °C
  • Melting Pt:251…255 °C
  • Flash Pt:>110 °C(230 °F)
  • Storage Temperature:Refrigerator
  • EINECS:257-530-2
Specifications for Product Family

Specification Test Results

  • Assay:
    98%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $108.83
    $108.83
    Each (10g)
       $108.83

    Supplier Number: J66437-09
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $629.79
    $629.79
    Each (100g)
       $629.79

    Supplier Number: J66437-22
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.