Order Entry
United States
ContactUsLinkComponent
Specpure® Viscosity Standard, Nominally 500Cst @ 40° and 52Cst @ 100°
Specpure® Viscosity Standard, Nominally 500Cst @ 40° and 52Cst @ 100°
Catalog # AA47155-AP
CAS Number:  
Specpure® Viscosity Standard, Nominally 500Cst @ 40° and 52Cst @ 100°
Catalog # AA47155-AP
Supplier Number:  047155-AP
CAS Number:  

Specifications

  • Size:
    500 ml
  • MDL No.:
    MFCD00084411
  • Physical Form:
    Liquid
  • Cat. No.:
    AA47155-AP
  • Supplier no.:
    47155-AP

Specifications

About this item

Viscosity standard, Specpure®, nominally 500cSt @ 40° and 52cSt @ 100°.

Physical Form: Liquid
MDL No.: MFCD00084411