Orders LinkContactUsLinkComponent
Dilution Bottles
Dilution Bottles
Dilution Bottles
Certificates
Dilution Bottles
3 Options Available Below
Product Details & Documents
GVS dilution bottles are pre-filled and sterile, available in 90 and 99 ml sizes.
  • Pre-filled for easy handling
  • Offered in 90 and 99 ml volumes for easy 1:10 and 1:100 dilutions
  • Compliance with standard methods
GVS pre-filled sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH (±0.2 pH) at 25 °C. They come in an easy open, flip-top, plastic container with a tamper-evident seal.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 10057-622
    Dilution bottle, Butterfield's buffer
    Dilution bottle, Butterfield's buffer
    90 ml
    Log in for availability
    Log in for availability
    Pack of 72 (90ml)
    Pack of 72 (90ml) - $247.70
    $247.70
    Pack of 72 (90ml)
       $247.70
    Supplier Number: 10498504
  • 10057-624
    Dilution bottle, Butterfield's buffer
    Dilution bottle, Butterfield's buffer
    99 ml
    Log in for availability
    Log in for availability
    Pack of 72 (99ml)
    Pack of 72 (99ml) - $240.37
    $240.37
    Pack of 72 (99ml)
       $240.37
    Supplier Number: 10498503
  • 10057-626
    Dilution bottle, Phosphate buffer Magnesium Chloride
    Dilution bottle, Phosphate buffer Magnesium Chloride
    99 ml
    Log in for availability
    Log in for availability
    Pack of 72 (99ml)
    Pack of 72 (99ml) - $237.84
    $237.84
    Pack of 72 (99ml)
       $237.84
    Supplier Number: 10498505