ContactUsLinkComponent
Pierce™ LAL Chromogenic Endotoxin Quantitation Kit, Thermo Scientific
Pierce™ LAL Chromogenic Endotoxin Quantitation Kit, Thermo Scientific
Catalog # PI88282
Supplier:  Avantor
CAS Number:  
undefined
Pierce™ LAL Chromogenic Endotoxin Quantitation Kit, Thermo Scientific
Catalog # PI88282
Supplier:  Avantor
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
About this item
The Pierce LAL Chromogenic Endotoxin Quantitation Kit measures the amount of endotoxin in a protein, peptide, or antibody sample using the Limulus Amebocyte Lysate (LAL) assay.
  • Sensitive - detect as little as 0.1 EU/ml
  • Fast - perform this assay in 30 minutes using a 96-well microplate
The endotoxin concentration in a sample is measured using the Pierce LAL Chromogenic Endotoxin Quantitation Kit via a chromogenic signal generated in the presence of endotoxins. Samples can be measured on a microplate absorbance reader at 405nm. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation and/or sepsis in tissue culture cells and animals injected with endotoxin contaminated proteins.
Delivery: Kit includes Limulus Amebocyte Lysate (LAL), E. coli endotoxin standard, chromogenic substrate and endotoxin-free water to prepare assay reagents.